3-[(2-Pyrrolidin-1-ylethyl)carbamoyl]benzeneboronic acid hydrochloride 98%

Lieferant: Apollo Scientific

OR7354-1G
APOSOR7354-1GEA 372 EUR
APOSOR7354-1G
3-[(2-Pyrrolidin-1-ylethyl)carbamoyl]benzeneboronic acid hydrochloride 98%
3-[(2-Pyrrolidin-1-ylethyl)carbamoyl]benzeneboronic acid hydrochloride
Formel: C₁₃H₁₉BN₂O₃·ClH
Molecular Weight: 298,574 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027257
CAS-Nummer: 957061-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR