4'-Chlorobiphenyl-4-carboxaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[1,1'-Biphenyl]-4-carboxaldehyde, 4'-chloro-, 4'-Chlorobiphenyl-4-carbaldehyde, 4'-Chloro-4-biphenylcarbaldehyde, 4'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde
OR7357-5G
APOSOR7357-5GEA
72.5
EUR
APOSOR7357-5G
4'-Chlorobiphenyl-4-carboxaldehyde 97%
4'-Chlorobiphenyl-4-carboxaldehyde
Formel:
C₁₃H₉ClO Molecular Weight: 216,67 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631911 CAS-Nummer: 80565-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...