3-Acetyl-2'-chlorobiphenyl
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2'-Chlorobiphenyl-3-yl)ethan-1-one, 3-(2-Chlorophenyl)acetophenone
OR7367-250MG
APOSOR7367-250MGEA
60.9
EUR
APOSOR7367-250MG
3-Acetyl-2'-chlorobiphenyl
3-Acetyl-2'-chlorobiphenyl
Formel:
C₁₄H₁₁ClO Molecular Weight: 230,69 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039058 CAS-Nummer: 675596-35-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...