4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid

Lieferant: Apollo Scientific
Achtung

OR7376-1G
APOSOR7376-1GEA 510 EUR
APOSOR7376-1G
4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid
4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid
Formel: C₁₆H₁₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00269588
CAS-Nummer: 857492-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR