2-(Fur-2-yl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

OR7396-100MG
APOSOR7396-100MGEA 102 EUR
APOSOR7396-100MG
2-(Fur-2-yl)benzaldehyde
2-(Fur-2-yl)benzaldehyde
Formel: C₁₁H₈O₂
Molecular Weight: 172.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03990475
CAS-Nummer: 16191-32-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR