2-(Fur-2-yl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

OR7399-1G OR7399-250MG OR7399-5G
APOSOR7399-1GEA 0 EUR
APOSOR7399-1G APOSOR7399-250MG APOSOR7399-5G
2-(Fur-2-yl)benzoic acid 97%
2-(Fur-2-yl)benzoic acid
Formel: C₁₁H₈O₃
Schmelzpunkt: 87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03990477
CAS-Nummer: 331942-47-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR