4-(Fur-2-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Carboxyphenyl)furan

OR7401-250MG
APOSOR7401-250MGEA 94.4 EUR
APOSOR7401-250MG
4-(Fur-2-yl)benzoic acid
4-(Fur-2-yl)benzoic acid
Formel: C₁₁H₈O₃
Molecular Weight: 188,179 g/mol
Schmelzpunkt: 231…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039073
CAS-Nummer: 35461-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR