3-(2-Furyl)benzonitril

Lieferant: Apollo Scientific
Achtung

OR7403-1G OR7403-250MG OR7403-50MG
APOSOR7403-1GEA 235 EUR
APOSOR7403-1G APOSOR7403-250MG APOSOR7403-50MG
3-(2-Furyl)benzonitril
3-(2-Furyl)benzonitril
Formel: C₁₁H₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD04039075
CAS-Nummer: 112598-77-3
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR