2-Chlor-5-cyan-3-pyridinboronsäure 96%

Lieferant: Apollo Scientific
Achtung

OR7415-250MG
APOSOR7415-250MGEA 306 EUR
APOSOR7415-250MG
2-Chlor-5-cyan-3-pyridinboronsäure 96%
2-Chloro-5-cyano-3-pyridineboronic acid
Formel: C₆H₄BClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09027243
CAS-Nummer: 957060-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR