3-[2-(Diethylamino)ethylcarbamoyl]benzeneboronic acid hydrochloride 97%
Lieferant: Apollo Scientific
OR7419-1G
APOSOR7419-1GEA
219
EUR
APOSOR7419-1G
3-[2-(Diethylamino)ethylcarbamoyl]benzeneboronic acid hydrochloride 97%
3-[2-(Diethylamino)ethylcarbamoyl]benzeneboronic acid hydrochloride
Formel:
C₁₃H₂₁BN₂O₃·HCl Molecular Weight: 300,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027256 CAS-Nummer: 957061-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...