α-Methyl-4-(methylsulphonyl)benzylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-(Methylsulphonyl)phenyl)ethanamine

OR7443-1G
APOSOR7443-1GEA 24.8 EUR
APOSOR7443-1G
α-Methyl-4-(methylsulphonyl)benzylamine
alpha-Methyl-4-(methylsulphonyl)benzylamine
Formel: C₉H₁₃NO₂S
Molecular Weight: 199,27 g/mol
Schmelzpunkt: 65…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319250
CAS-Nummer: 387350-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR