2-Amino-3'-methoxybiphenyl hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Methoxyphenyl)aniline hydrochloride, 3'-Methoxy-[1,1'-biphenyl]-2-amine hydrochloride

OR7444-500MG
APOSOR7444-500MGEA 26.1 EUR
APOSOR7444-500MG
2-Amino-3'-methoxybiphenyl hydrochloride
2-Amino-3'-methoxybiphenyl hydrochloride
Formel: C₁₃H₁₃NO·ClH
Molecular Weight: 235,709 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739419
CAS-Nummer: 1082749-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR