3'-Methoxy-[1,1'-biphenyl]-4-amine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3'-Methoxybiphenyl-4-ylamine hydrochloride, 4-(3-Methoxyphenyl)aniline hydrochloride

OR7450-1G
APOSOR7450-1GEA 43.5 EUR
APOSOR7450-1G
3'-Methoxy-[1,1'-biphenyl]-4-amine hydrochloride
3'-Methoxy-[1,1'-biphenyl]-4-amine hydrochloride
Formel: C₁₃H₁₃NO.HCl
Molecular Weight: 235,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772428
CAS-Nummer: 207287-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR