3'-Methoxybiphenyl-3-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR7464-250MG
APOSOR7464-250MGEA
23.2
EUR
APOSOR7464-250MG
3'-Methoxybiphenyl-3-carbonsäure
3'-Methoxybiphenyl-3-carbonsäure
Formel:
C₁₄H₁₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424598 CAS-Nummer: 168618-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...