4'-Methoxybiphenyl-2-carbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Methoxy[1,1'-biphenyl]-2-carboxylic acid, 4'-Methoxy-[1,1'-biphenyl]-2-carboxylic acid

OR7466-250MG OR7466-100MG
APOSOR7466-250MGEA 58.2 EUR
APOSOR7466-250MG APOSOR7466-100MG
4'-Methoxybiphenyl-2-carbonsäure
4'-Methoxybiphenyl-2-carbonsäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03426469
CAS-Nummer: 18110-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR