1-(4'-Methoxy-[1,1'-biphenyl]-2-yl)ethan-1-one

Lieferant: Apollo Scientific
Achtung

OR7471-1G
APOSOR7471-1GEA 571 EUR
APOSOR7471-1G
1-(4'-Methoxy-[1,1'-biphenyl]-2-yl)ethan-1-one
1-(4'-Methoxy-[1,1'-biphenyl]-2-yl)ethan-1-one
Formel: C₁₅H₁₄O₂
Molecular Weight: 226,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039102
CAS-Nummer: 192863-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR