4'-(3-Methoxyphenyl)acetophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3'-Methoxy[1,1'-biphenyl]-4-yl)ethan-1-one

OR7476-1G
APOSOR7476-1GEA 256 EUR
APOSOR7476-1G
4'-(3-Methoxyphenyl)acetophenon
4'-(3-Methoxyphenyl)acetophenon
Formel: C₁₅H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04039105
CAS-Nummer: 76650-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR