5-(3-Methoxyphenyl)-2-furoic acid
Lieferant: Apollo Scientific
|
Gefahr
|
OR7479-250MG
APOSOR7479-250MGEA
36.4
EUR
APOSOR7479-250MG
5-(3-Methoxyphenyl)-2-furoic acid
5-(3-Methoxyphenyl)-2-furoic acid
Formel:
C₁₂H₁₀O₄ Molecular Weight: 218,205 g/mol Schmelzpunkt: 169…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05022489 CAS-Nummer: 54022-96-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...