Methyl 3'-amino-[1,1'-biphenyl]-3-carboxylate hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-[3-(Methoxycarbonyl)phenyl]aniline hydrochloride, 3'-Amino-3-(methoxycarbonyl)biphenyl hydrochloride

OR7484-100MG
APOSOR7484-100MGEA 21.9 EUR
APOSOR7484-100MG
Methyl 3'-amino-[1,1'-biphenyl]-3-carboxylate hydrochloride 98%
Methyl 3'-amino-[1,1'-biphenyl]-3-carboxylate hydrochloride
Formel: C₁₄H₁₃NO₂·ClH
Molecular Weight: 263,719 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366482
CAS-Nummer: 343985-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR