2'-Methyl[1,1'-biphenyl]-2-amine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR7490-500MG
APOSOR7490-500MGEA 212 EUR
APOSOR7490-500MG
2'-Methyl[1,1'-biphenyl]-2-amine hydrochloride
2'-Methyl[1,1'-biphenyl]-2-amine hydrochloride
Formel: C₁₃H₁₃N·ClH
Molecular Weight: 219,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739417
CAS-Nummer: 1203-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR