4-Amino-2'-methylbiphenyl hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(2-Methylphenyl)aniline hydrochloride, 2'-Methyl-[1,1'-biphenyl]-4-amine hydrochloride

OR7496-1G
APOSOR7496-1GEA 174 EUR
APOSOR7496-1G
4-Amino-2'-methylbiphenyl hydrochloride
4-Amino-2'-methylbiphenyl hydrochloride
Formel: C₁₃H₁₃N·ClH
Molecular Weight: 219,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739442
CAS-Nummer: 811842-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR