4-Benzylanilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Aminodiphenylmethane
OR7498-25G
APOSOR7498-25GEA
84.1
EUR
APOSOR7498-25G
4-Benzylanilin 98%
4-Benzylanilin
Formel:
C₁₃H₁₃N Molecular Weight: 183,25 g/mol Siedepunkt: 142…144 °C/1mmHg Schmelzpunkt: 35…38 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00047861 CAS-Nummer: 1135-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...