4-(3-Bromopropoxy)benzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (4-(3-Bromopropoxy)phenyl)boronic acid

OR7501-1G
APOSOR7501-1GEA 234 EUR
APOSOR7501-1G
4-(3-Bromopropoxy)benzeneboronic acid 98%
4-(3-Bromopropoxy)benzeneboronic acid
Formel: C₉H₁₂BBrO₃
Molecular Weight: 258,905 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258751
CAS-Nummer: 957034-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR