2-Methoxy-4,6-dimethylnicotinonitrile

Lieferant: Apollo Scientific

Synonyme: 4,6-Dimethyl-2-methoxynicotinonitrile

OR7502-1G
APOSOR7502-1GEA 52.2 EUR
APOSOR7502-1G
2-Methoxy-4,6-dimethylnicotinonitrile
2-Methoxy-4,6-dimethylnicotinonitrile
Formel: C₉H₁₀N₂O
Molecular Weight: 162,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00124716
CAS-Nummer: 65515-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR