3'-Methylbiphenyl-4-carboxaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3'-Methyl-[1,1'-biphenyl]-4-carbaldehyde

OR7510-1G
APOSOR7510-1GEA 62.5 EUR
APOSOR7510-1G
3'-Methylbiphenyl-4-carboxaldehyde 95%
3'-Methylbiphenyl-4-carboxaldehyde
Formel: C₁₄H₁₂O
Molecular Weight: 196,249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424618
CAS-Nummer: 400744-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR