2'-Methylbiphenyl-3-carbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2'-Methyl-[1,1'-biphenyl]-3-carboxylic acid

OR7514-500MG
APOSOR7514-500MGEA 21.9 EUR
APOSOR7514-500MG
2'-Methylbiphenyl-3-carbonsäure
2'-Methylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424589
CAS-Nummer: 168618-44-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR