2'-Methylbiphenyl-4-carbonsäure

Lieferant: Apollo Scientific
Achtung

OR7515-1G OR7515-250MG
APOSOR7515-1GEA 92.8 EUR
APOSOR7515-1G APOSOR7515-250MG
2'-Methylbiphenyl-4-carbonsäure
2'-Methylbiphenyl-4-carbonsäure
Formel: C₁₄H₁₂O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 5748-43-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR