3'-Methyl-[1,1'-biphenyl]-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Carboxy-3'-methylbiphenyl, 2-(3-Methylphenyl)benzoic acid

OR7516-1G OR7516-5G
APOSOR7516-1GEA 125 EUR
APOSOR7516-1G APOSOR7516-5G
3'-Methyl-[1,1'-biphenyl]-2-carboxylic acid
3'-Methyl-[1,1'-biphenyl]-2-carboxylic acid
Formel: C₁₄H₁₂O₂
Schmelzpunkt: 94…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039113
CAS-Nummer: 107412-71-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR