3'-Methylbiphenyl-3-carbonsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3'-Methyl[1,1'-biphenyl]-3-carboxylic acid
OR7517-1G
APOSOR7517-1GEA
26.1
EUR
APOSOR7517-1G
3'-Methylbiphenyl-3-carbonsäure 97%
3'-Methylbiphenyl-3-carbonsäure
Formel:
C₁₄H₁₂O₂ Molecular Weight: 212.25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00558892 CAS-Nummer: 158619-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...