3'-Methylbiphenyl-4-carbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3'-Methyl-[1,1'-biphenyl]-4-carboxylic acid

OR7518-1G
APOSOR7518-1GEA 37.7 EUR
APOSOR7518-1G
3'-Methylbiphenyl-4-carbonsäure
3'-Methylbiphenyl-4-carbonsäure
Formel: C₁₄H₁₂O₂
Molecular Weight: 212.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424590
CAS-Nummer: 5728-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR