2,2'-Methylenebis(6-tert-butyl-p-cresol) 99+%

Lieferant: Apollo Scientific

OR75243-500G OR75243-2.5KG
APOSOR75243-500GEA 89 EUR
APOSOR75243-500G APOSOR75243-2.5KG
2,2'-Methylenebis(6-tert-butyl-p-cresol) 99+%
2,2'-Methylenebis(6-tert-butyl-p-cresol)
Formel: C₂₃H₃₂O₂
MDL: MFCD00043641
CAS-Nummer: 119-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR