2-(5-Methylfur-2-yl)aniline hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR7535-500MG
APOSOR7535-500MGEA
98.9
EUR
APOSOR7535-500MG
2-(5-Methylfur-2-yl)aniline hydrochloride
2-(5-Methylfur-2-yl)aniline hydrochloride
Formel:
C₁₁H₁₁NO·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD01191726 CAS-Nummer: 329187-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...