2-Amino-3,5-dibrompyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dibrompyridin-2-amin, 3,5-Dibrom-2-pyridylamin
OR7544-5G
APOSOR7544-5GEA
23.4
EUR
APOSOR7544-5G
2-Amino-3,5-dibrompyridin
2-Amino-3,5-dibrompyridin
Formel:
C₅H₄Br₂N₂ Molecular Weight: 251,91 g/mol Schmelzpunkt: 102…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038041 CAS-Nummer: 35486-42-1 EINECS: 252-590-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...