Methyl-2-(2-furyl)benzoate

Lieferant: Apollo Scientific

OR7545-1G OR7545-250MG OR7545-500MG OR7545-5G
APOSOR7545-1GEA 261 EUR
APOSOR7545-1G APOSOR7545-250MG APOSOR7545-500MG APOSOR7545-5G
Methyl-2-(2-furyl)benzoate
Methyl-2-(2-furyl)benzoate
Formel: C₁₂H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04039126
CAS-Nummer: 39732-01-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR