4-(5-Methylfur-2-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR7555-250MG
APOSOR7555-250MGEA 27.7 EUR
APOSOR7555-250MG
4-(5-Methylfur-2-yl)benzoic acid
4-(5-Methylfur-2-yl)benzoic acid
Formel: C₁₂H₁₀O₃
Schmelzpunkt: 226 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03032492
CAS-Nummer: 53782-63-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR