Methyl 3-(3-hydroxyprop-1-yn-1-yl)benzoate

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-[3-(Methoxycarbonyl)phenyl]prop-2-yn-1-ol

OR7568-250MG
APOSOR7568-250MGEA 43.5 EUR
APOSOR7568-250MG
Methyl 3-(3-hydroxyprop-1-yn-1-yl)benzoate
Methyl 3-(3-hydroxyprop-1-yn-1-yl)benzoate
Formel: C₁₁H₁₀O₃
Molecular Weight: 190,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039139
CAS-Nummer: 121114-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR