5-(4-Methylphenyl)-2-furoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(4-Methylphenyl)furan-2-carboxylic acid, 2-Carboxy-5-(4-methylphenyl)furan, 4-(5-Carboxyfur-2-yl)toluene, 5-(p-Tolyl)furan-2-carboxylic acid

OR7577-1G
APOSOR7577-1GEA 58.2 EUR
APOSOR7577-1G
5-(4-Methylphenyl)-2-furoic acid
5-(4-Methylphenyl)-2-furoic acid
Formel: C₁₂H₁₀O₃
Molecular Weight: 202,206 g/mol
Schmelzpunkt: 179…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00448781
CAS-Nummer: 52938-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR