3-(Thien-2-yl)aniline 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(3-Aminophenyl)thiophene, 3-(Thiophen-2-yl)aniline
OR7584-250MG
APOSOR7584-250MGEA
81.2
EUR
APOSOR7584-250MG
3-(Thien-2-yl)aniline 96%
3-(Thien-2-yl)aniline
Formel:
C₁₀H₉NS Molecular Weight: 175,25 g/mol Schmelzpunkt: 246…247 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05022493 CAS-Nummer: 92057-12-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...