4-(Thien-2-yl)aniline hydrochloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR7585-100MG
APOSOR7585-100MGEA
241
EUR
APOSOR7585-100MG
4-(Thien-2-yl)aniline hydrochloride
4-(Thien-2-yl)aniline hydrochloride
Formel:
C₁₀H₉NS·ClH Schmelzpunkt: 185…187 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00136945 CAS-Nummer: 1172363-16-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...