1-Dimethylamino-2-nitroethylene 98%

Lieferant: Apollo Scientific
Achtung

OR7588-10G OR7588-100G OR7588-25G
APOSOR7588-10GEA 161 EUR
APOSOR7588-10G APOSOR7588-100G APOSOR7588-25G
1-Dimethylamino-2-nitroethylene 98%
1-Dimethylamino-2-nitroethylene
Formel: C₄H₈N₂O₂
Schmelzpunkt: 103…105 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00051519
CAS-Nummer: 1190-92-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR