2-(Thien-3-yl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Thiophen-3-yl)benzaldehyde

OR7590-1G
APOSOR7590-1GEA 227 EUR
APOSOR7590-1G
2-(Thien-3-yl)benzaldehyde
2-(Thien-3-yl)benzaldehyde
Formel: C₁₁H₈OS
Molecular Weight: 188,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039149
CAS-Nummer: 99902-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR