3-(Thien-3-yl)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(3-Carboxyphenyl)thiophene
OR7594-1G
APOSOR7594-1GEA
116
EUR
APOSOR7594-1G
3-(Thien-3-yl)benzoic acid 97%
3-(Thien-3-yl)benzoic acid
Formel:
C₁₁H₈O₂S Molecular Weight: 204,245 g/mol Schmelzpunkt: 177…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039153 CAS-Nummer: 20608-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...