Ethyl-6-bromimidazo[1,2-a]pyridin-2-carboxylat 98%

Lieferant: Apollo Scientific
Achtung

OR7600-5G
APOSOR7600-5GEA 91.5 EUR
APOSOR7600-5G
Ethyl-6-bromimidazo[1,2-a]pyridin-2-carboxylat 98%
Ethyl-6-bromimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₀H₉BrN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD06659034
CAS-Nummer: 67625-37-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR