BRN 0043822
Lieferant: Apollo Scientific
|
Achtung
|
OR7600T-1G
APOSOR7600T-1GEA
46.4
EUR
APOSOR7600T-1G
BRN 0043822
BRN 0043822
Formel:
C₁₉H₂₆O₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269929 CAS-Nummer: 18685-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...