1-[4-(2-Thienyl)phenyl]ethanone

Lieferant: Apollo Scientific

Synonyme: 1-(4-(Thiophen-2-yl)phenyl)ethanone

OR7603-1G
APOSOR7603-1GEA 409 EUR
APOSOR7603-1G
1-[4-(2-Thienyl)phenyl]ethanone
1-[4-(2-Thienyl)phenyl]ethanone
Formel: C₁₂H₁₀OS
Molecular Weight: 202,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039158
CAS-Nummer: 35294-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR