4-Hydroxybenzamidin Hydrochlorid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Hydroxyphenylamidin Hydrochlorid, 4-Hydroxybenzoesäureamidin Hydrochlorid

OR7609-1G
APOSOR7609-1GEA 21.9 EUR
APOSOR7609-1G
4-Hydroxybenzamidin Hydrochlorid 95+%
4-Hydroxybenzamidin Hydrochlorid
Formel: C₇H₉ClN₂O
Molecular Weight: 172,61 g/mol
Schmelzpunkt: 223…226 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00136405
CAS-Nummer: 38148-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR