Methyl (4-hydroxypyrimidin-2-ylsulphanyl)acetate

Lieferant: Apollo Scientific
Achtung

OR7610-1G
APOSOR7610-1GEA 165 EUR
APOSOR7610-1G
Methyl (4-hydroxypyrimidin-2-ylsulphanyl)acetate
Methyl (4-hydroxypyrimidin-2-ylsulphanyl)acetate
Formel: C₇H₈N₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00185401
CAS-Nummer: 79361-42-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR