2-(1,8-Naphthyridin-2-yl)phenol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7617-1G
APOSOR7617-1GEA
328
EUR
APOSOR7617-1G
2-(1,8-Naphthyridin-2-yl)phenol 95+%
2-(1,8-Naphthyridin-2-yl)phenol
Formel:
C₁₄H₁₀N₂O Schmelzpunkt: 193…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00203050 CAS-Nummer: 65182-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...