2-(4-Bromophenyl)malondialdehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR7621-1G
APOSOR7621-1GEA
0
EUR
APOSOR7621-1G
2-(4-Bromophenyl)malondialdehyde 95%
2-(4-Bromophenyl)malondialdehyde
Formel:
C₉H₇BrO₂ Schmelzpunkt: 157…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02261895 CAS-Nummer: 709648-68-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...