4-Chloro-2-(methylsulphonyl)benzaldehyde 95+%

Lieferant: Apollo Scientific
Achtung

OR7625-1G
APOSOR7625-1GEA 244 EUR
APOSOR7625-1G
4-Chloro-2-(methylsulphonyl)benzaldehyde 95+%
4-Chloro-2-(methylsulphonyl)benzaldehyde
Formel: C₈H₇ClO₃S
Molecular Weight: 218.66 g/mol
Schmelzpunkt: 112…114 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD04037946
CAS-Nummer: 849035-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR